Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAGUAAUCGCGGUUUUUGUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548h-5p | MIMAT0005928 |
| Callithrix jacchus | cja-miR-548f | MIMAT0039576 |
| Pan troglodytes | ptr-miR-548h | MIMAT0008222 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-548h-1 |
| Stem-loop Accession # | MI0006411 |
| Stem-loop Sequence | UCUGUCCAUUAGGUGGGUGCAAAAGUAAUCGCGGUUUUUGUCAUUACUUUUAAUGGUAAAAACUGGAAUUACUUUUGCACUGACCUAAUAUUAAGCCAGAUA |
| Chromosome Location | Chr. 14 - 64095024 - 64095125 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005928
miRBase ID: hsa-miR-548h-5p Mature miRNA Sequence: AAAAGUAAUCGCGGUUUUUGUC Chromosome Location: Chr. 8 - 27048853 - 27048963 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-548h-2 |
| Stem-loop Accession # | MI0006412 |
| Stem-loop Sequence | GUAUUAGGUUGGUGCAAAAGUAAUCGCGGUUUUUGUCAUUACUUUCAAUGGCAAACACCACAAUUACUUUUGCACCAACCUAAUAUAA |
| Chromosome Location | Chr. 16 - 11306440 - 11306527 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005928
miRBase ID: hsa-miR-548h-5p Mature miRNA Sequence: AAAAGUAAUCGCGGUUUUUGUC Chromosome Location: Chr. 8 - 27048853 - 27048963 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-548h-3 |
| Stem-loop Accession # | MI0006413 |
| Stem-loop Sequence | UCUGAUUCUGCAUGUAUUAGGUUGGUGCAAAAGUAAUCGCGGUUUUUGUCAUUGAAAGUAAUAGCAAAAACUGCAAUUACUUUUGCACCAACCUAAAAGUAGUCACUGUCUUCAGAUA |
| Chromosome Location | Chr. 17 - 13543529 - 13543646 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005928
miRBase ID: hsa-miR-548h-5p Mature miRNA Sequence: AAAAGUAAUCGCGGUUUUUGUC Chromosome Location: Chr. 8 - 27048853 - 27048963 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-548h-4 |
| Stem-loop Accession # | MI0006414 |
| Stem-loop Sequence | GCUAUUAGGUUGGUGCAAAAGUAAUCGCGGUUUUUGUCAUUACUUUAAUUACUUUACGUUUCAUUAAUGACAAAAACCGCAAUUACUUUUGCACCAACCUAAUACUUGCUA |
| Chromosome Location | Chr. 8 - 27048853 - 27048963 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005928
miRBase ID: hsa-miR-548h-5p Mature miRNA Sequence: AAAAGUAAUCGCGGUUUUUGUC Chromosome Location: Chr. 8 - 27048853 - 27048963 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-548h-5 |
| Stem-loop Accession # | MI0016751 |
| Stem-loop Sequence | ACAAAAGUAAUCGCGGUUUUUGUCAUUACUUUUAACUGUAAAAACCACGGUUGCUUUUGC |
| Chromosome Location | Chr. 6 - 131792172 - 131792231 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005928
miRBase ID: hsa-miR-548h-5p Mature miRNA Sequence: AAAAGUAAUCGCGGUUUUUGUC Chromosome Location: Chr. 8 - 27048853 - 27048963 [-] on Build GRCh38 |
| Stem-loop ID | cja-mir-548f |
| Stem-loop Accession # | MI0032109 |
| Stem-loop Sequence | UAUUAGGUUGGUGCAAAAGUAAUCGCGGUUUUUGUCAUAAAACCAUUACUUUCGUGGCAAGAACCUCCAUUACUUUUGCACCAACCUAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039576
miRBase ID: cja-miR-548f Mature miRNA Sequence: AAAAGUAAUCGCGGUUUUUGUC Chromosome Location: NA |
| Stem-loop ID | ptr-mir-548h |
| Stem-loop Accession # | MI0008752 |
| Stem-loop Sequence | UCUGUCCAUUAGGUGGGUGCAAAAGUAAUCGCGGUUUUUGUCAUUACUUUUAAUGGUAAAAACUGGAAUUACUUUUGCACUGACCUAAUAUUAAGCCAUAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008222
miRBase ID: ptr-miR-548h Mature miRNA Sequence: AAAAGUAAUCGCGGUUUUUGUC Chromosome Location: NA |
More Information
Related Products

Genomic Map