Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGUGCAUCCUUUUAGAGUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-519a-3p | MIMAT0002869 |
| Gorilla gorilla | ggo-miR-519a-3p | MIMAT0036440 |
| Gorilla gorilla | ggo-miR-519a | MIMAT0036444 |
| Pan troglodytes | ptr-miR-519a | MIMAT0008192 |
| Pan troglodytes | ptr-miR-519f-3p | MIMAT0036457 |
| Pongo pygmaeus | ppy-miR-519a | MIMAT0015960 |
| View More Rows | ||
| Rhesus monkey | mml-miR-517b | MIMAT0036446 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map