Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. X - 146028093 - 146028093 [-] on 146028093 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-891a-3p | MIMAT0026717 |
| Human | hsa-miR-891a-5p | MIMAT0004902 |
Stem-loop Details
Human

Genomic Map