Assay Details
Amplicon Length: | 107 |
Chromosome Location: | Chr. 9 - 93595061 - 93595061 [-] on 93595061 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548au-5p | MIMAT0022291 |
| Human | hsa-miR-548au-3p | MIMAT0022292 |
Stem-loop Details
Human

Genomic Map