Assay Details
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | UAUUGCACAUUCACCGGCCUGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila melanogaster | dme-miR-311-3p | MIMAT0000403 |
Drosophila erecta | der-miR-311a-3p | MIMAT0008489 |
Drosophila sechellia | dse-miR-311-3p | MIMAT0008799 |
Drosophila simulans | dsi-miR-311-3p | MIMAT0008857 |
Drosophila yakuba | dya-miR-311a-3p | MIMAT0009091 |
Stem-loop Details
Drosophila melanogaster
Drosophila erecta
Drosophila sechellia
Drosophila simulans
Drosophila yakuba

More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-311 (v16)
Drosophila erecta: der-miR-311a (v21)
Drosophila sechellia: dse-miR-311 (v21)
Drosophila simulans: dsi-miR-311 (v21)
Drosophila yakuba: dya-miR-311a (v21)
Drosophila erecta: der-miR-311a (v21)
Drosophila sechellia: dse-miR-311 (v21)
Drosophila simulans: dsi-miR-311 (v21)
Drosophila yakuba: dya-miR-311a (v21)
