Assay Details
Mature miRNA Sequence: | AACUGGCCUACAAAGUCCCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chrysemys picta | cpi-miR-193a-3p | MIMAT0037810 |
Gorilla gorilla | ggo-miR-193a | MIMAT0024174 |
Monodelphis domestica | mdo-miR-193a-3p | MIMAT0004123 |
Stem-loop Details
Chrysemys picta
Gorilla gorilla
Monodelphis domestica

More Information
Related Products
miRBase Alias:
Monodelphis domestica: mdo-miR-193 (v19)
