Assay Details
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | AUCAACAAACAUUUAUUGUGUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-545-3p | MIMAT0003807 |
Pig | ssc-miR-545-3p | MIMAT0020594 |
Stem-loop Details
Bovine
Pig

More Information
Related Products
miRBase Alias:
Bos taurus: bta-miR-545 (v18)
Staphylococcus schleiferi: ssc-miR-545 (v18)
Staphylococcus schleiferi: ssc-miR-545 (v18)
