Assay Details
Mature miRNA Sequence: | CCAGUUAGGGUUUAAUGGUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dictyostelium discoideum | ddi-miR-1177 | MIMAT0005532 |
Stem-loop Details
Dictyostelium discoideum

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ddi-miR-1177 |
---|---|
miRBase Accession Number: | MI0006245 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CCAGUUAGGGUUUAAUGGUUC |
Species: | Dictyostelium discoideum |
Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | CCAGUUAGGGUUUAAUGGUUC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Dictyostelium discoideum | ddi-miR-1177 | MIMAT0005532 |