Assay Details
Gene Family ID: | MIPF0000564, mir-1421 |
Mature miRNA Sequence: | UGCAUCACGUGUGACCUGCCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ornithorhynchus anatinus | oan-miR-1421k-3p | MIMAT0009179 |
Stem-loop Details
Ornithorhynchus anatinus

More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-1421k-2-3p (v19)
Ochrobactrum anthropi: oan-miR-1421k-3* (v18)
Ochrobactrum anthropi: oan-miR-1421k-2* (v18)
Ochrobactrum anthropi: oan-miR-1421k-3-3p (v19)
Ochrobactrum anthropi: oan-miR-1421k-3* (v18)
Ochrobactrum anthropi: oan-miR-1421k-2* (v18)
Ochrobactrum anthropi: oan-miR-1421k-3-3p (v19)
