Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | AACAUCACUGCAAGUCUGUGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-499-3p | MIMAT0006950 |
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-499* (v18)

Genomic Map