Assay Details
Mature miRNA Sequence: | GCUGCAGAGCCGUGCGCGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ornithorhynchus anatinus | oan-miR-1354-3p | MIMAT0006948 |
Stem-loop Details
Ornithorhynchus anatinus

More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-1354* (v18)
