Assay Details
Gene Family ID: | MIPF0001913, mir-2041 |
Mature miRNA Sequence: | AUUUUGCUCUUACGUUUCACU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Nematostella vectensis | nve-miR-2041a-5p | MIMAT0009804 |
Stem-loop Details
Nematostella vectensis

More Information
Related Products
miRBase Alias:
Nematostella vectensis: nve-miR-2041-5p (v20)
Nematostella vectensis: nve-miR-2041* (v18)
Nematostella vectensis: nve-miR-2041* (v18)
