Assay Details
Gene Family ID: | MIPF0000938, mir-2355 |
Mature miRNA Sequence: | CCCAGAUACAAUGGACAAUAUGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2355-5p | MIMAT0011891 |
Stem-loop Details
Bovine

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bta-miR-2355-5p |
---|---|
miRBase Accession Number: | MI0011384 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CCCAGAUACAAUGGACAAUAUGC |
Species: | Bovine |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000938, mir-2355 |
Mature miRNA Sequence: | CCCAGAUACAAUGGACAAUAUGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-2355-5p | MIMAT0011891 |