Assay Details
Gene Family ID: | MIPF0000772, mir-2208 |
Mature miRNA Sequence: | AUGCAGUUUCUGGUAUACUUCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Caenorhabditis elegans | cel-miR-2208a-3p | MIMAT0011428 |
Stem-loop Details
Caenorhabditis elegans

More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-2208a* (v17)
