Assay Details
Mature miRNA Sequence: | AUGCAGAAGUGCACGGAAACAGCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-2131-5p | MIMAT0011207 |
Columba livia | cli-miR-2131-5p | MIMAT0038707 |
Stem-loop Details
Chicken
Columba livia
