Assay Details
Mature miRNA Sequence: | AAGUGGCAUUUGAUAAGCCAUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Caenorhabditis elegans | cel-miR-2212-3p | MIMAT0011440 |
Stem-loop Details
Caenorhabditis elegans

More Information
Related Products
miRBase Alias:
Caenorhabditis elegans: cel-miR-2212* (v17)
