Assay Details
Mature miRNA Sequence: | GCAAGUCAAUAAGAUUAUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila pseudoobscura | dps-miR-2562-5p | MIMAT0012386 |
Stem-loop Details
Drosophila pseudoobscura

More Information
Related Products
miRBase Alias:
Derxomyces pseudoschimicola: dps-miR-2562* (v18)
