Assay Details
Gene Family ID: | MIPF0001009, mir-1822 |
Mature miRNA Sequence: | CGAGCUGCCCGGGGAUAGACUCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Caenorhabditis briggsae | cbr-miR-1822 | MIMAT0011481 |
Stem-loop Details
Caenorhabditis briggsae
