Assay Details
Gene Family ID: | MIPF0000548, mir-422 |
Mature miRNA Sequence: | ACUGGACUGAGGGUCAGAAGGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pongo pygmaeus | ppy-miR-422a | MIMAT0015881 |
Stem-loop Details
Pongo pygmaeus

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ppy-miR-422a |
---|---|
miRBase Accession Number: | MI0014938 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUGGACUGAGGGUCAGAAGGC |
Species: | Pongo pygmaeus |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000548, mir-422 |
Mature miRNA Sequence: | ACUGGACUGAGGGUCAGAAGGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Pongo pygmaeus | ppy-miR-422a | MIMAT0015881 |