Assay Details
Gene Family ID: | MIPF0000010, MIR159 |
Mature miRNA Sequence: | ACUGGAUGACGCGGGAGCUAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Aegilops tauschii | ata-miR319-3p | MIMAT0037245 |
| Rice | osa-miR319a-3p | MIMAT0020915 |
Stem-loop Details
Aegilops tauschii
Rice

Genomic Map