Assay Details
Gene Family ID: | MIPF0000078, mir-187 |
Mature miRNA Sequence: | GGCUGCAACACAGGACAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Oryzias latipes | ola-miR-187 | MIMAT0022596 |
Stem-loop Details
Oryzias latipes

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ola-miR-187 |
---|---|
miRBase Accession Number: | MI0019478 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GGCUGCAACACAGGACAU |
Species: | Oryzias latipes |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000078, mir-187 |
Mature miRNA Sequence: | GGCUGCAACACAGGACAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Oryzias latipes | ola-miR-187 | MIMAT0022596 |