Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | AGUGACAUCACAUAUACGGCGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-489 | MIMAT0025375 |
Stem-loop Details
Pig
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ssc-miR-489 |
|---|---|
| miRBase Accession Number: | MI0022150 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGUGACAUCACAUAUACGGCGG |
| Species: | Pig |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | AGUGACAUCACAUAUACGGCGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-489 | MIMAT0025375 |