Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | AGAUAUGUUUGAUAUUCUUGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Heliconius melpomene | hme-miR-190 | MIMAT0024944 |
Stem-loop Details
Heliconius melpomene
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hme-miR-190 |
|---|---|
| miRBase Accession Number: | MI0021739 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGAUAUGUUUGAUAUUCUUGGU |
| Species: | Heliconius melpomene |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | AGAUAUGUUUGAUAUUCUUGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Heliconius melpomene | hme-miR-190 | MIMAT0024944 |