Assay Details
Gene Family ID: | MIPF0000360, mir-551 |
Mature miRNA Sequence: | GAAACCAAGUGUGGGUGUGGCCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Salmo salar | ssa-miR-551-5p | MIMAT0032663 |
Stem-loop Details
Salmo salar

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ssa-miR-551-5p |
---|---|
miRBase Accession Number: | MI0026720 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GAAACCAAGUGUGGGUGUGGCCU |
Species: | Salmo salar |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000360, mir-551 |
Mature miRNA Sequence: | GAAACCAAGUGUGGGUGUGGCCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Salmo salar | ssa-miR-551-5p | MIMAT0032663 |