Assay Details
Mature miRNA Sequence: | AGACCCUGGUCUGCACUCUAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pteropus alecto | pal-miR-504-5p | MIMAT0040011 |
Stem-loop Details
Pteropus alecto
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | pal-miR-504-5p |
|---|---|
| miRBase Accession Number: | MI0032447 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGACCCUGGUCUGCACUCUAU |
| Species: | Pteropus alecto |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | AGACCCUGGUCUGCACUCUAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pteropus alecto | pal-miR-504-5p | MIMAT0040011 |