Assay Details
Gene Family ID: | MIPF0000431, mir-744 |
Mature miRNA Sequence: | CUGUUGCCACUAACCUCAACCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-744-3p | MIMAT0004946 |
| Mouse | mmu-miR-744-3p | MIMAT0004820 |
Stem-loop Details
Human
Mouse
More Information
Related Products
miRBase Alias:
Human: hsa-miR-744* (v17)
Mouse: mmu-miR-744* (v17)
Mouse: mmu-miR-744* (v17)

Genomic Map