Assay Details
Gene Family ID: | MIPF0000519, mir-563 |
Mature miRNA Sequence: | AGGCUGACAUACAUUUCCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rhesus monkey | mml-miR-563 | MIMAT0006429 |
Stem-loop Details
Rhesus monkey

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mml-miR-563 |
---|---|
miRBase Accession Number: | MI0007836 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGGCUGACAUACAUUUCCC |
Species: | Rhesus monkey |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000519, mir-563 |
Mature miRNA Sequence: | AGGCUGACAUACAUUUCCC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rhesus monkey | mml-miR-563 | MIMAT0006429 |