Assay Details
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | UGUUCGUUCGGCUCGCGUUAUCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Branchiostoma floridae | bfl-miR-375 | MIMAT0009501 |
Stem-loop Details
Branchiostoma floridae
