Assay Details
Mature miRNA Sequence: | AACUGGCCCGUCAAGUCCCUCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Capitella teleta | cte-miR-193 | MIMAT0009533 |
Stem-loop Details
Capitella teleta
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cte-miR-193 |
|---|---|
| miRBase Accession Number: | MI0010077 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACUGGCCCGUCAAGUCCCUCC |
| Species: | Capitella teleta |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | AACUGGCCCGUCAAGUCCCUCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Capitella teleta | cte-miR-193 | MIMAT0009533 |