Assay Details
Mature miRNA Sequence: | UACAUGUAUAGUAUUAUUUGCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-2172-3p | MIMAT0011272 |
Stem-loop Details
Schmidtea mediterranea
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | sme-miR-2172-3p |
|---|---|
| miRBase Accession Number: | MI0010815 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACAUGUAUAGUAUUAUUUGCCU |
| Species: | Schmidtea mediterranea |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UACAUGUAUAGUAUUAUUUGCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-2172-3p | MIMAT0011272 |