Assay Details
Mature miRNA Sequence: | AGGGCAGGCAUAGCUAAGUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-2206-5p | MIMAT0011402 |
Stem-loop Details
Schmidtea mediterranea
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | sme-miR-2206-5p |
|---|---|
| miRBase Accession Number: | MI0010939 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGGCAGGCAUAGCUAAGUG |
| Species: | Schmidtea mediterranea |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | AGGGCAGGCAUAGCUAAGUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-2206-5p | MIMAT0011402 |