Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | GUGACAUCAUAUGUACGGCUGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Anolis carolinensis | aca-miR-489-3p | MIMAT0021975 |
Taeniopygia guttata | tgu-miR-489-3p | MIMAT0014618 |
Stem-loop Details
Anolis carolinensis
Taeniopygia guttata

More Information
Related Products
miRBase Alias:
Trabulsiella guamensis ATCC 49490: tgu-miR-489 (v19)
