Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | AGAUAUGUUUGAUAUUCUUGGUUGUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Nasonia vitripennis | nvi-miR-190 | MIMAT0015673 |
Stem-loop Details
Nasonia vitripennis
