Assay Details
Gene Family ID: | MIPF0000975, mir-2494 |
Mature miRNA Sequence: | GUAAGUGCACCAGAUUGGGAAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Drosophila simulans | dsi-miR-2494-5p | MIMAT0012521 |
Stem-loop Details
Drosophila simulans

More Information
Related Products
miRBase Alias:
Drosophila simulans: dsi-miR-2494 (v18)
