Assay Details
Mature miRNA Sequence: | UCAGUAACAUGGGUUCGAACCUCG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Petromyzon marinus | pma-miR-459 | MIMAT0019555 |
Stem-loop Details
Petromyzon marinus

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | pma-miR-459 |
---|---|
miRBase Accession Number: | MI0017137 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UCAGUAACAUGGGUUCGAACCUCG |
Species: | Petromyzon marinus |
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UCAGUAACAUGGGUUCGAACCUCG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Petromyzon marinus | pma-miR-459 | MIMAT0019555 |