Assay Details
Gene Family ID: | MIPF0000635, mir-1329 |
Mature miRNA Sequence: | UACAGUGAUCAGGUUACGAUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Anolis carolinensis | aca-miR-1329-5p | MIMAT0021747 |
Stem-loop Details
Anolis carolinensis

More Information
Related Products
miRBase Alias:
Aeromonas caviae Ae398: aca-miR-1329 (v18)
