Assay Details
Gene Family ID: | MIPF0000915, mir-2253 |
Mature miRNA Sequence: | CAUGGCACUGCUGAAACUUCGAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pristionchus pacificus | ppc-miR-2253d-3p | MIMAT0033004 |
Stem-loop Details
Pristionchus pacificus

Genomic Map