Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | CGAAUGUUGCUCGGUGAACCCCUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Eptesicus fuscus | efu-miR-409 | MIMAT0035041 |
Stem-loop Details
Eptesicus fuscus

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | efu-miR-409 |
---|---|
miRBase Accession Number: | MI0028726 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CGAAUGUUGCUCGGUGAACCCCUU |
Species: | Eptesicus fuscus |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | CGAAUGUUGCUCGGUGAACCCCUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Eptesicus fuscus | efu-miR-409 | MIMAT0035041 |