Assay Details
Gene Family ID: | MIPF0000102, MIR408 |
Mature miRNA Sequence: | ACAGGGAACAAGCAGAGCAUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR408-5p | MIMAT0031915 |
Stem-loop Details
Arabidopsis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ath-miR408-5p |
|---|---|
| miRBase Accession Number: | MI0001080 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACAGGGAACAAGCAGAGCAUG |
| Species: | Arabidopsis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000102, MIR408 |
Mature miRNA Sequence: | ACAGGGAACAAGCAGAGCAUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR408-5p | MIMAT0031915 |