Assay Details
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | ACGUUGAGCCACACGCACAAUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Salmo salar | ssa-miR-375-5p | MIMAT0032625 |
Stem-loop Details
Salmo salar

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | ssa-miR-375-5p |
---|---|
miRBase Accession Number: | MI0026695 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACGUUGAGCCACACGCACAAUA |
Species: | Salmo salar |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | ACGUUGAGCCACACGCACAAUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Salmo salar | ssa-miR-375-5p | MIMAT0032625 |