Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CCUCUACAGGGAAGCGCUUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-519a-2-5p | MIMAT0037327 |
| Human | hsa-miR-520b-5p | MIMAT0037325 |
Stem-loop Details
Human

Genomic Map