Assay Details
Mature miRNA Sequence: | UUCACAAAGCCCAUACACUUUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Guinea pig | cpo-miR-350-3p | MIMAT0047187 |
Stem-loop Details
Guinea pig
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cpo-miR-350-3p |
|---|---|
| miRBase Accession Number: | MI0038741 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCACAAAGCCCAUACACUUUCA |
| Species: | Guinea pig |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UUCACAAAGCCCAUACACUUUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Guinea pig | cpo-miR-350-3p | MIMAT0047187 |