Assay Details
Amplicon Length: | 51 |
Chromosome Location: | Chr. 19 - 6264443 - 6264443 [-] on 6264443 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-194-2-3p | MIMAT0017073 |
| Mouse | mmu-miR-194-5p | MIMAT0000224 |
Stem-loop Details
Mouse

Genomic Map