Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 19 - 29135579 - 29135579 [-] on 29135579 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-101b-5p | MIMAT0017046 |
| Mouse | mmu-miR-101b-3p | MIMAT0000616 |
Stem-loop Details
Mouse

Genomic Map