Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 1 - 185313180 - 185313180 [-] on 185313180 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-194-5p | MIMAT0000224 |
| Mouse | mmu-miR-194-1-3p | MIMAT0016999 |
Stem-loop Details
Mouse

Genomic Map