Assay Details
Amplicon Length: | 86 |
Chromosome Location: | Chr. 2 - 80628406 - 80628406 [-] on 80628406 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-684 | MIMAT0003462 |
Stem-loop Details
Mouse
