Assay Details
Amplicon Length: | 71 |
Chromosome Location: | Chr. 4 - 124743387 - 124743387 [+] on 124743387 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-698-5p | MIMAT0022930 |
| Mouse | mmu-miR-698-3p | MIMAT0003488 |
Stem-loop Details
Mouse

Genomic Map