Assay Details
Amplicon Length: | 81 |
Chromosome Location: | Chr. 8 - 84011111 - 84011111 [-] on 84011111 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1199-3p | MIMAT0017333 |
| Mouse | mmu-miR-1199-5p | MIMAT0005860 |
Stem-loop Details
Mouse

Genomic Map