Assay Details
Gene Family ID: | MIPF0000055, mir-194 |
Mature miRNA Sequence: | CCAGUGGGGCUGCUGUUAUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-194-3p | MIMAT0017148 |
| Pig | ssc-miR-194a-3p | MIMAT0037465 |
Stem-loop Details
Rat
Pig
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-194-3p |
|---|---|
| miRBase Accession Number: | MI0000938 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCAGUGGGGCUGCUGUUAUCU |
| Species: | Rat, Pig |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000055, mir-194 |
Mature miRNA Sequence: | CCAGUGGGGCUGCUGUUAUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-194-3p | MIMAT0017148 |
| Pig | ssc-miR-194a-3p | MIMAT0037465 |