Assay Details
Amplicon Length: | 71 |
Chromosome Location: | Chr. 8 - 120378802 - 120378802 [+] on 120378802 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-128-3p | MIMAT0000834 |
| Rat | rno-miR-128-2-5p | MIMAT0017119 |
Stem-loop Details
Rat

Genomic Map